Human recombinant GUCY2C (from HEK293 cells), biotin
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Biotin
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His, Avitag™
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Gene ID:P25092-1
- Protein synonyms:GUCY2C|hSTAR|STA receptor|GC-C|STAR|GUC2C
- Protein/peptide name:GUCY2C
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:49,6 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4.
- Shipping temperature:RT
Specifications
Frequently Bought Together