Human recombinant integrin alpha 4 beta 1 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Natalizumab at 1 μg/ml (100 μl/well) can bind Human ITGA4 and ITGB1 Heterodimer Protein, His Tag and Tag Free (Cat. No. ACRBIT1-H52W1) with a linear range of 1 - 16 ng/ml (QC tested).
- Gene ID:P13612-1 (ITGA4) and P05556-1 (ITGB1)
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:Integrin alpha 4 beta 1|ITGA4&ITGB1
- Protein/peptide name:Integrin alpha 4 beta 1
- Purity:>90%
- Molecular weight:111,4 kDa (ITGA4) and 83,7 kDa (ITGB1)
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together