Human recombinant IL-7 R alpha (from HEK293)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised ActiveMax® Human IL-7, Tag Free (Cat. No. ACRBIL7-H4219) at 2 μg/ml (100 μl/well) can bind Human IL-7 R alpha, Mouse IgG2a Fc Tag (Cat. No. ACRBIL7-H5258) with a linear range of 4-31 ng/ml (QC tested).; Measured by its binding ability in a functional ELISA. Biotinylated Human TSLP (R127A, R130A), His,Avitag (Cat. No. ACRBTSP-H82E0) immobilised at 2 μg/ml (100 μl/well) via precoated 5 μg/ml (100 μl/well) of Human TSLP R, Fc Tag (Cat. No. ACRBTSR-H525a), can bind can bind Human IL-7 R alpha, Mouse IgG2a Fc Tag (Cat. No. ACRBIL7-H5258) with a linear range of 1 - 20 ng/ml (Routinely tested).; Captured Human IL-7 R alpha, Mouse IgG2a Fc Tag (Cat. No. ACRBIL7-H5258) on CM5 chip via anti-mouse antibodies surface can bind ActiveMax® Human IL-7, Tag Free (Cat. No. ACRBIL7-H4219) with an affinity constant of 13,3 nM as determined in a SPR assay (Biacore T200) (Routinely tested).
- Gene ID:P16871-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:IL7Ra|CD127
- Protein/peptide name:IL-7 R alpha
- Purity:>90%
- Molecular weight:52,0 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Frequently Bought Together