Human recombinant IL-3 R alpha (from HEK293)
Nome del fornitore: ACRO Biosystems
Certificati
Informazioni su questo articolo
Specifiche
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation. Immobilised Human IL-3 R alpha, His Tag (SPR verified) (Cat. No. ACRBILA-H52H6) on CM5 Chip via anti-His antibody can bind Human IL-3, Tag Free with an affinity constant of 224 nM as determined in SPR assay (Biacore T200) (QC tested).; Loaded Human IL-3 R alpha, His Tag (SPR verified) (Cat. No. ACRBILA-H52H6) on NTA Biosensor, can bind Human IL-3, Tag Free with an affinity constant of 370 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P26951-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:IL3RX|IL3RA|CD123 antigen|IL-3Ra|IL3RY|CD123|IL-3 R alpha|hIL3Ra|MGC34174|hIL-3Ra|IL3RAY|IL3R|IL-3R-alpha
- Protein/peptide name:IL-3 R alpha
- Purity:>95%
- Molecular weight:35,0 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifiche
Frequently Bought Together