Human recombinant IL-1 Rrp2 (from HEK293)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Gene ID:Q9HB29-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:IL1RL2|IL-1RL2|IL-36R|IL-1Rrp2|IL1Rrp2|IL-1 R6|IL1RRP2|IL-1 Rrp2
- Protein/peptide name:IL-1 Rrp2
- Purity:>90%
- Molecular weight:37,8 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Frequently Bought Together