Human recombinant CD30 ligand (from HEK293)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 0,1 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Human CD30, His Tag (Cat. No. CD0-H5229) at 2 μg/ml (100 μl/well) can bind Human CD30 Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDL-H525b) with a linear range of 1 - 25 ng/ml (QC tested). Loaded Biotinylated Human CD30, Avitag,His Tag (Cat. No. CD0-H82E6) on SA Biosensor, can bind Human CD30 Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDL-H525b) with an affinity constant of 114 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). FACS analysis shows that Human CD30 Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDL-H525b) can bind to K562-CD30 cells surface CD30. The concentration of Human CD30 Ligand is 0,1 μg/ml (Routinely tested). FACS analysis shows that the binding of Human CD30 Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDL-H525b) to K562-CD30 cells surface CD30 was inhibited by increasing concentration of neutralising anti-CD30 antibody. The concentration of Human CD30 Ligand used is 0,1 μg/ml. The IC50 is 0,2064 μg/ml (Routinely tested).
- Gene ID:P32971-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:CD30 antigen ligand|CD30-L|TNFSF8|CD30 Ligand|CD30LG|CD153|CD30L|CD153 antigen
- Protein/peptide name:CD30 Ligand
- Purity:>95%
- Molecular weight:46,5 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder, RT
Frequently Bought Together