Human recombinant BAFFR (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Human BAFF, His Tag (Cat. No. BAF-H5248) at 5 μg/ml (100 μl/well) can bind Human BAFFR, Fc Tag (Cat. No. BAR-H5257) with a linear range of 2 - 31 ng/ml (QC tested). Immobilised Biotinylated Human BAFF, Avitag,Fc Tag (active trimer) (MALS verified) (Cat. No. BAF-H82F3) on SA Chip can bind Human BAFFR, Fc Tag (Cat. No. BAR-H5257) with an affinity constant of 26,7 nM as determined in a SPR assay (Biacore T200) (Routinely tested).
- Gene ID:Q96RJ3-1
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:CVID4|BAFFR|BAFF-R|BROMIX|CD268|TNFRSF13C|prolixin
- Protein/peptide name:BAFFR
- Purity:>95%
- Molecular weight:33,2 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder, RT
- Carrier-free:Y
Specifications
Frequently Bought Together