Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant ANGPTL7 (from HEK293)
Human recombinant ANGPTL7 (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant ANGPTL7 (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Gene ID:
    O43827-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    CDT6|ANGPTL7
  • Protein/peptide name:
    ANGPTL7
  • Purity:
    >90%
  • Molecular weight:
    64,1 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder, RT

Specifications

Frequently Bought Together