Human recombinant nectin-3 (from HEK293)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7,5, Normally trehalose is added as protectant before Lyophilisation,; Loaded Human Nectin-3, Fc Tag (Cat, No, PV3-H5255) on Protein A Biosensor, can bind Human TIGIT, His Tag (Cat, No, TIT-H52H3) with an affinity constant of 0,732 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:NP_056295.1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:CD113|Nectin-3|PVRL3
- Protein/peptide name:Nectin-3
- Purity:>95%
- Molecular weight:64,3 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5, Normally trehalose is added as protectant before Lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Frequently Bought Together