Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant Mesothelin (from HEK293)
Human recombinant Mesothelin (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human recombinant Mesothelin (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4, Normally trehalose is added as protectant before Lyophilisation,; Measured by its binding ability in a functional ELISA, Immobilised Anti-Human MSLN MAb at 0,5 μg/ml (100 μl/well) can bind Biotinylated Human Mesothelin, Fc,Avitag (Cat, No, MSN-H82F6) with a linear range of 0,2-6 ng/ml (QC tested).
  • Gene ID:
    Q13421-3(R309P)
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    MPF|MSLN|Mesothelin
  • Protein/peptide name:
    Mesothelin
  • Purity:
    >95%
  • Molecular weight:
    60,4 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4, Normally trehalose is added as protectant before Lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together