Human recombinant LILRA5 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Human ANGPTL7, His Tag (Cat. No. AN7-H52H5) at 5 μg/ml(100 μl/well) can bind Biotinylated Human LILRA5, His,Avitag (Cat. No. LI5-H82E1) with a linear range of 1.25-20 μg/ml(QC tested).
- Gene ID:A6NI73-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:ILT11|LIR9|LILRA5|LILRB7|CD85f
- Protein/peptide name:LILRA5
- Purity:>95%
- Molecular weight:28,9 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4, Normally trehalose is added as protectant before Lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together