Human recombinant LIGHT (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 0,1 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before Lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Human HVEM, His Tag (Cat. No. HVM-H52E9) at 5 μg/ml(100 μl/well) can bind Human LIGHT, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. LIT-H5256) with a linear range of 0.3-5 ng/ml(QC tested).; Loaded Human DcR3, Fc Tag (Cat. No. TNB-H5255) on AHC Biosensor, can bind Human LIGHT, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. LIT-H5256) with an affinity constant of 1.64 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:O43557-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:HVEM-L|CD258|LTg|TR2|LIGHT|HVEML|CD258 antigen|TNFSF14
- Protein/peptide name:Light
- Purity:>95%
- Molecular weight:45,1 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5, Normally trehalose is added as protectant before Lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together