Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chloro-4-fluoro-1H-indole-2-carboxylic acid
5-Chloro-4-fluoro-1H-indole-2-carboxylic acid 97%
5-Chloro-4-fluoro-1H-indole-2-carboxylic acid 97%
5-Chloro-4-fluoro-1H-indole-2-carboxylic acid 97%
APOSPC908743-1G
:
Apollo Scientific
:
5-Chloro-4-fluoro-1H-indole-2-carboxylic acid 97%
APOSPC908743-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
213.593 g/mol
:
Ambient
:
186446-26-4
:
C₉H₅ClFNO₂
:
MFCD09835295