Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Fluoro-1H-indole-5-carboxylic acid
4-Fluoro-1H-indole-5-carboxylic acid ≥98%
4-Fluoro-1H-indole-5-carboxylic acid ≥98%
APOSPC540008-250MG
:
Apollo Scientific
:
4-Fluoro-1H-indole-5-carboxylic acid ≥98%
APOSPC540008-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
179.148 g/mol
:
Refrigerator
:
908600-72-6
:
C₉H₆FNO₂
:
MFCD12924428