Order Entry
Switzerland
ContactUsLinkComponent
6-Chloro-3-(difluoromethyl)-1H-indole
6-Chloro-3-(difluoromethyl)-1H-indole
Katalognr. APOSPC520111-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Chloro-3-(difluoromethyl)-1H-indole
Katalognr. APOSPC520111-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :201.6 g/mol
Storage Temperature:Ambient
Formula:C₉H₆ClF₂N
MDL Number:MFCD29767594