3-(Difluoromethyl)-6-methyl-1H-indole
Katalognr. APOSPC520099-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:181.182 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₉F₂N
MDL Number:MFCD29767582