Order Entry
Switzerland
ContactUsLinkComponent
3-(Difluoromethyl)-5-methyl-1H-indole-2-carboxylic acid
3-(Difluoromethyl)-5-methyl-1H-indole-2-carboxylic acid
Catalog # APOSPC520075-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(Difluoromethyl)-5-methyl-1H-indole-2-carboxylic acid
Catalog # APOSPC520075-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :225.191 g/mol
Storage Temperature:Ambient
CAS Number:2092578-19-1
Formula:C₁₁H₉F₂NO₂
MDL Number:MFCD29767559