6-[2-(Trifluoromethoxy)phenyl]-3-pyridinecarbaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:898405-31-7
:
Formula:C₁₃H₈F₃NO₂
MDL Number:MFCD04115434
:267.203 g/mol
Storage Temperature:Ambient
Frequently Bought Together