
2-[3-(Trifluoromethyl)phenyl]pyrrolidine
Catalog # APOSPC450017-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:215.215 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:109086-17-1
Formula:C₁₁H₁₂F₃N
MDL Number:MFCD02663523