4-Bromo-3-(trifluoromethyl)pyridine hydrobromide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1354425-60-7
:
Formula:C₆H₃BrF₃N·BrH
MDL Number:MFCD19690886
:306.906 g/mol
Melting Pt:197…209 °C
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together