2-(4-Chlorophenyl)-2'-fluoroacetophenone
Catalog # APOSPC421016-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:248.68 g/mol
Storage Temperature:Ambient
CAS Number:1183199-32-7
Formula:C₁₄H₁₀ClFO
MDL Number:MFCD16350637