3-(Trifluoromethyl)cyclobutan-1-amine hydrochloride
Catalog # APOSPC411274-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:175.58 g/mol
Storage Temperature:Ambient
CAS Number:1803601-06-0
Formula:C₅H₈F₃N·ClH
MDL Number:MFCD28714563