Order Entry
Switzerland
ContactUsLinkComponent
1-[5-(Trifluoromethyl)pyridin-2-yl]-1H-pyrazol-3-amine hydrochloride
1-[5-(Trifluoromethyl)pyridin-2-yl]-1H-pyrazol-3-amine hydrochloride
Catalog # APOSPC410095-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[5-(Trifluoromethyl)pyridin-2-yl]-1H-pyrazol-3-amine hydrochloride
Catalog # APOSPC410095-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :264.635 g/mol
Storage Temperature:Ambient
Formula:C₉H₇F₃N₄·ClH
MDL Number:MFCD25371156