4-[4-(Difluoromethoxy)phenyl]-2-hydrazinylpyrimidine
Catalog # APOSPC410045-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:252.22 g/mol
Storage Temperature:Ambient
CAS Number:932240-78-3
Formula:C₁₁H₁₀F₂N₄O
MDL Number:MFCD08556206