1-(4-Fluoro-benzyl)-piperidine-4-carboxylic acid
Catalog # APOSPC408316-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:237.27 g/mol
Storage Temperature:Ambient
CAS Number:193538-25-9
Formula:C₁₃H₁₆FNO₂
MDL Number:MFCD03371459