
3-[4-(Trifluoromethyl)phenyl]oxetan-3-ol
Catalog # APOSPC405502-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:218.172 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:26755-29-3
Formula:C₁₀H₉F₃O₂
MDL Number:MFCD27664912