Order Entry
Switzerland
ContactUsLinkComponent
4-Fluoro-5-iodo-1H-indole
4-Fluoro-5-iodo-1H-indole
Catalog # APOSPC402055-250MG
Supplier:  Apollo Scientific
CAS Number:  
4-Fluoro-5-iodo-1H-indole
Catalog # APOSPC402055-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :261.035 g/mol
Storage Temperature:Ambient
CAS Number:1240113-40-9
Formula:C₈H₅FIN
MDL Number:MFCD22628143