Order Entry
Switzerland
ContactUsLinkComponent
tert-Butyl 4-[3-(4-fluorophenoxy)-2-hydroxypropyl]piperazine-1-carboxylate
tert-Butyl 4-[3-(4-fluorophenoxy)-2-hydroxypropyl]piperazine-1-carboxylate
Catalog # APOSPC300527-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl 4-[3-(4-fluorophenoxy)-2-hydroxypropyl]piperazine-1-carboxylate
Catalog # APOSPC300527-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :354.416 g/mol
Storage Temperature:Ambient
CAS Number:874801-05-5
Formula:C₁₈H₂₇FN₂O₄
MDL Number:MFCD05664816