Order Entry
Switzerland
ContactUsLinkComponent
1-(3,3,3-Trifluoro-2-hydroxypropyl)piperidin-4-ol, tech
1-(3,3,3-Trifluoro-2-hydroxypropyl)piperidin-4-ol, tech
Catalog # APOSPC28178-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(3,3,3-Trifluoro-2-hydroxypropyl)piperidin-4-ol, tech
Catalog # APOSPC28178-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :213.197 g/mol
Storage Temperature:Ambient
CAS Number:478081-38-8
Formula:C₈H₁₄F₃NO₂
MDL Number:MFCD01871721