Order Entry
Switzerland
ContactUsLinkComponent
6-Methyl-3-[3-(trifluoromethyl)phenyl]-1,2,4-triazin-5-ol, tech
6-Methyl-3-[3-(trifluoromethyl)phenyl]-1,2,4-triazin-5-ol, tech
Catalog # APOSPC28173-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Methyl-3-[3-(trifluoromethyl)phenyl]-1,2,4-triazin-5-ol, tech
Catalog # APOSPC28173-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :255.196 g/mol
Storage Temperature:Ambient
CAS Number:339102-74-8
Formula:C₁₁H₈F₃N₃O
MDL Number:MFCD00665260