Order Entry
Switzerland
ContactUsLinkComponent
Ethyl 4-(3,3,3-trifluoro-2-hydroxypropyl)piperazine-1-carboxylate, tech
Ethyl 4-(3,3,3-trifluoro-2-hydroxypropyl)piperazine-1-carboxylate, tech
Catalog # APOSPC28160-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 4-(3,3,3-trifluoro-2-hydroxypropyl)piperazine-1-carboxylate, tech
Catalog # APOSPC28160-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :270.249 g/mol
Storage Temperature:Ambient
CAS Number:866135-53-7
Formula:C₁₀H₁₇F₃N₂O₃
MDL Number:MFCD05256288