Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(3E)-4-(2,5-Difluorophenyl)but-3-en-2-one
(3E)-4-(2,5-Difluorophenyl)but-3-en-2-one, tech
(3E)-4-(2,5-Difluorophenyl)but-3-en-2-one, tech
(3E)-4-(2,5-Difluorophenyl)but-3-en-2-one, tech
APOSPC28129-1G
:
Apollo Scientific
:
(3E)-4-(2,5-Difluorophenyl)but-3-en-2-one, tech
APOSPC28129-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
182.167 g/mol
:
Ambient
:
1571074-31-1
:
C₁₀H₈F₂O
:
MFCD03305731