3-Fluoroquinoxalin-2-ol, tech
Catalog # APOSPC28070-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:164.137 g/mol
Storage Temperature:Ambient
CAS Number:477889-54-6
Formula:C₈H₅FN₂O
MDL Number:MFCD01815123