Order Entry
Switzerland
ContactUsLinkComponent
(2Z)-2-{[2-(Trifluoromethyl)phenyl]methylidene}-1-azabicyclo[2.2.2]octan-3-ol, tech
(2Z)-2-{[2-(Trifluoromethyl)phenyl]methylidene}-1-azabicyclo[2.2.2]octan-3-ol, tech
Catalog # APOSPC27956-1G
Supplier:  Apollo Scientific
CAS Number:  
(2Z)-2-{[2-(Trifluoromethyl)phenyl]methylidene}-1-azabicyclo[2.2.2]octan-3-ol, tech
Catalog # APOSPC27956-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :283.289 g/mol
Storage Temperature:Ambient
CAS Number:477848-11-6
Formula:C₁₅H₁₆F₃NO
MDL Number:MFCD02081948