

2-[4-(Bromomethyl)phenyl]-1,1,1,3,3,3-hexafluoropropan-2-ol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:202134-57-4
:
Formula:C₁₀H₇BrF₆O
MDL Number:MFCD09998077
:337.056 g/mol
UN:1760
ADR:8,III
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together