2-(4-(Trifluoromethyl)phenyl)propan-2-amine hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:15996-89-1
:
Formula:C₁₀H₁₂F₃N·ClH
MDL Number:MFCD14708149
:239.665 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together