Methyl 2-[(3,3,3-trifluoro-2-hydroxypropyl)sulphanyl]benzenecarboxylate
Catalog # APOSPC10279-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:280.263 g/mol
Boiling Pt:158…160 °C (1 mm Hg)
Storage Temperature:Ambient
CAS Number:385383-55-1
Formula:C₁₁H₁₁F₃O₃S
MDL Number:MFCD03001288