
Ethyl 2-(trifluoromethyl)-1,8-naphthyridine-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:252959-76-5
:
Formula:C₁₂H₉F₃N₂O₂
MDL Number:MFCD00202892
Melting Pt:125 °C
:270.207 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together