Order Entry
Switzerland
ContactUsLinkComponent
1,1,1-Trifluoro-3-[4-(3,3,3-trifluoro-2-hydroxypropyl)homopiperazin-1-yl]propan-2-ol
1,1,1-Trifluoro-3-[4-(3,3,3-trifluoro-2-hydroxypropyl)homopiperazin-1-yl]propan-2-ol
Catalog # APOSPC10115-500MG
Supplier:  Apollo Scientific
CAS Number:  
1,1,1-Trifluoro-3-[4-(3,3,3-trifluoro-2-hydroxypropyl)homopiperazin-1-yl]propan-2-ol
Catalog # APOSPC10115-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :324.263 g/mol
Melting Pt:77…79 °C
Storage Temperature:Ambient
CAS Number:477847-09-9
Formula:C₁₁H₁₈F₆N₂O₂
MDL Number:MFCD02082052