Order Entry
Switzerland
ContactUsLinkComponent
6-Isopropoxypyridin-3-ol 98%
6-Isopropoxypyridin-3-ol 98%
Catalog # APOSOR957496-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Isopropoxypyridin-3-ol 98%
Catalog # APOSOR957496-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :153.178 g/mol
Storage Temperature:Ambient
CAS Number:903886-71-5
Formula:C₈H₁₁NO₂
MDL Number:MFCD16997398