Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Phenyl-2-(pyrazin-2-yl)pyrimidin-4-ol
6-Phenyl-2-(pyrazin-2-yl)pyrimidin-4-ol
6-Phenyl-2-(pyrazin-2-yl)pyrimidin-4-ol
APOSOR9554-50MG
:
Apollo Scientific
:
6-Phenyl-2-(pyrazin-2-yl)pyrimidin-4-ol
APOSOR9554-50MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
50 mg
Seq:
2370
:
:
250.255 g/mol
:
GHS07
:
Ambient
:
874605-79-5
:
C₁₄H₁₀N₄O
:
MFCD00665788