
3-Methylazetidin-3-ol 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:256931-54-1
:
Formula:C₄H₉NO
MDL Number:MFCD10565787
:87.12 g/mol
Storage Temperature:Refrigerator
UN:1759
ADR:8,III
:GHS05
Frequently Bought Together