Order Entry
Switzerland
ContactUsLinkComponent
5-Azaspiro[2.5]octan-8-one hydrochloride
5-Azaspiro[2.5]octan-8-one hydrochloride
Catalog # APOSOR935112-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Azaspiro[2.5]octan-8-one hydrochloride
Catalog # APOSOR935112-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :161.629 g/mol
Storage Temperature:Ambient
CAS Number:1780166-60-0
Formula:C₇H₁₁NO·ClH
MDL Number:MFCD28501958