Order Entry
Switzerland
ContactUsLinkComponent
1-Isopropylpiperidin-4-ol 95%
1-Isopropylpiperidin-4-ol 95%
Catalog # APOSOR909302-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Isopropylpiperidin-4-ol 95%
Catalog # APOSOR909302-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :143.227 g/mol
Storage Temperature:Ambient
CAS Number:5570-78-5
Formula:C₈H₁₇NO
MDL Number:MFCD00101977