Order Entry
Switzerland
ContactUsLinkComponent
6,7-Dimethyl-1H-indole-2-carboxylic acid 95%
6,7-Dimethyl-1H-indole-2-carboxylic acid 95%
Catalog # APOSOR905679-1G
Supplier:  Apollo Scientific
CAS Number:  
6,7-Dimethyl-1H-indole-2-carboxylic acid 95%
Catalog # APOSOR905679-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :189.211 g/mol
Storage Temperature:Ambient
CAS Number:383132-15-8
Formula:C₁₁H₁₁NO₂
MDL Number:MFCD02664418