5-(Ethylsulphonyl)-2-methoxyaniline
Catalog # APOSOR8344-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:215.269 g/mol
Melting Pt:103…107 °C
EINECS:226-272-2
:GHS07
Storage Temperature:Refrigerator
CAS Number:5339-62-8
Formula:C₉H₁₃NO₃S
MDL Number:MFCD00025083