2-Bromo-3-methoxyaniline
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:112970-44-2
:
Formula:C₇H₈BrNO
MDL Number:MFCD07787450
ADR:6.1,III
:202.049 g/mol
UN:2811
:GHS06
:GHS07
Storage Temperature:Ambient
Frequently Bought Together