2-[2-(1-Chlorocyclopropyl)-3-(2-chlorophenyl)-2-hydroxypropyl]-1,2-dihydro-3H-1,2,4-triazole-3-thione 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:178928-70-6
:
Formula:C₁₄H₁₅Cl₂N₃OS
MDL Number:MFCD08273847
:344.259 g/mol
Storage Temperature:Ambient
Frequently Bought Together