
5,7-Dimethyl-1,2,3,4-tetrahydro-1,8-naphthyridine
Catalog # APOSOR55297-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:162.232 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:65541-95-9
Formula:C₁₀H₁₄N₂
MDL Number:MFCD13705330