Order Entry
Switzerland
ContactUsLinkComponent
Pent-4-en-2-amine hydrochloride 95%
Pent-4-en-2-amine hydrochloride 95%
Catalog # APOSOR55255-250MG
Supplier:  Apollo Scientific
CAS Number:  
Pent-4-en-2-amine hydrochloride 95%
Catalog # APOSOR55255-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590
  • Environmentally Preferable:

Specifications

About this item

 :
 :121.608 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1315366-25-6
Formula:C₅H₁₁N·ClH
MDL Number:MFCD19382008